4-Bromo-5-fluoro-2-methylaniline
Supplier: Apollo Scientific
Warning
PC500260-5G PC500260-1G PC500260-25G
APOSPC500260-5GEA 37.7 EUR
APOSPC500260-5G APOSPC500260-1G APOSPC500260-25G
4-Bromo-5-fluoro-2-methylaniline
4-Bromo-5-fluoro-2-methylaniline
Formula: C₇H₇BrFN
MW: 204.0400 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD04973753
CAS Number: 52723-82-7