Methyl-2-amino-6-fluoro-3-nitrobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-amino-6-fluoro-3-nitrobenzoate
PC500279-1G PC500279-25G PC500279-5G
APOSPC500279-1GEA 33.5 EUR
APOSPC500279-1G APOSPC500279-25G APOSPC500279-5G
Methyl-2-amino-6-fluoro-3-nitrobenzoate
Methyl-2-amino-6-fluoro-3-nitrobenzoate
Formula: C₈H₇FN₂O₄
MW: 214.15 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD09839221
CAS Number: 346691-23-4