4-Bromo-2,6-difluorobenzotrifluoride 99%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Bromo-1,3-difluoro-2-(trifluoromethyl)benzene
PC500419-1G PC500419-25G PC500419-5G
APOSPC500419-1GEA 21.9 EUR
APOSPC500419-1G APOSPC500419-25G APOSPC500419-5G
4-Bromo-2,6-difluorobenzotrifluoride 99%
4-Bromo-2,6-difluorobenzotrifluoride
Formula: C₇H₂BrF₅
MW: 260.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631616
CAS Number: 156243-64-0