Lithium hexafluorosilicate 98%
Supplier: Apollo Scientific
Danger
PC5005-25G PC5005-5G
APOSPC5005-25GEA 232 EUR
APOSPC5005-25G APOSPC5005-5G
Lithium hexafluorosilicate 98%
Lithium hexafluorosilicate
Formula: F₆Li₂Si
MW: 155.96 g/mol
Density: 2.01 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049473
CAS Number: 17347-95-4
EINECS: 241-372-6
UN: 2856
ADR: 6.1 III