4'-Fluoro-3'-(trifluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
PC500523-10G PC500523-1G PC500523-5G
APOSPC500523-10GEA 412 EUR
APOSPC500523-10G APOSPC500523-1G APOSPC500523-5G
4'-Fluoro-3'-(trifluoromethoxy)acetophenone 98%
4'-Fluoro-3'-(trifluoromethoxy)acetophenone
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660260
CAS Number: 886501-44-6