2-Fluoro-4-(methylsulphonyl)benzoic acid
Supplier: Apollo Scientific
Warning
PC501372-1G PC501372-5G
APOSPC501372-1GEA 77 EUR
APOSPC501372-1G APOSPC501372-5G
2-Fluoro-4-(methylsulphonyl)benzoic acid
2-Fluoro-4-(methylsulphonyl)benzoic acid
Formula: C₈H₇FO₄S
Storage Temperature: Ambient
MDL Number: MFCD11615492
CAS Number: 142994-04-5