1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 4-Fluoro-5-methyl-2-nitroanisole
PC501455-1G PC501455-5G
APOSPC501455-1GEA 139 EUR
APOSPC501455-1G APOSPC501455-5G
1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene
1-Fluoro-4-methoxy-2-methyl-5-nitrobenzene
Formula: C₈H₈FNO₃
MW: 185.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18157656
CAS Number: 182880-71-3