4-Bromo-2-chloro-5-fluoroaniline 95%
Supplier: Apollo Scientific
PC501469-1G PC501469-5G
APOSPC501469-1GEA 45.2 EUR
APOSPC501469-1G APOSPC501469-5G
4-Bromo-2-chloro-5-fluoroaniline 95%
4-Bromo-2-chloro-5-fluoroaniline
Formula: C₆H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD09878155
CAS Number: 1000572-63-3