4-Bromo-2,5-difluorobenzoic acid
Supplier: Apollo Scientific
Warning
PC501855-1G PC501855-25G PC501855-5G
APOSPC501855-1GEA 21.9 EUR
APOSPC501855-1G APOSPC501855-25G APOSPC501855-5G
4-Bromo-2,5-difluorobenzoic acid
4-Bromo-2,5-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MW: 237.0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00844105
CAS Number: 28314-82-1