Methyl-5-bromo-2,3-difluorobenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 5-bromo-2,3-difluorobenzoate
PC501961-1G PC501961-250MG PC501961-5G
APOSPC501961-1GEA 105 EUR
APOSPC501961-1G APOSPC501961-250MG APOSPC501961-5G
Methyl-5-bromo-2,3-difluorobenzoate
Methyl 5-bromo-2,3-difluorobenzoate
Formula: C₈H₅BrF₂O₂
MW: 251.0249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026006
CAS Number: 1150163-69-1