2-Fluoro-4-nitro-1-(trifluoromethyl)benzene
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4-nitrobenzotrifluoride
PC50207-1G PC50207-25G PC50207-5G
APOSPC50207-1GEA 87.3 EUR
APOSPC50207-1G APOSPC50207-25G APOSPC50207-5G
2-Fluoro-4-nitro-1-(trifluoromethyl)benzene
2-Fluoro-4-nitro-1-(trifluoromethyl)benzene
Formula: C₇H₃F₄NO₂
MW: 209.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782060
CAS Number: 69411-67-2
UN: 2810
ADR: 6.1,III