Methyl-2-chloro-5-fluorobenzoate
Supplier: Apollo Scientific
Synonyms: 2-Chloro-5-fluorobenzoic acid methyl ester
PC502676-25G PC502676-5G
APOSPC502676-25GEA 524 EUR
APOSPC502676-25G APOSPC502676-5G
Methyl-2-chloro-5-fluorobenzoate
Methyl 2-chloro-5-fluorobenzoate
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204207
CAS Number: 647020-63-1