Di-tert-butylsilyl-bis(trifluoromethanesulphonate)
Supplier: Apollo Scientific
Danger
PC5037-25G PC5037-5G PC5037-1G
APOSPC5037-25GEA 335 EUR
APOSPC5037-25G APOSPC5037-5G APOSPC5037-1G
Di-tert-butylsilyl-bis(trifluoromethanesulphonate)
Di-tert-butylsilyl-bis(trifluoromethanesulphonate)
Formula: C₁₀H₁₈F₆O₆S₂Si
MW: 440.44 g/mol
Boiling Pt: 263 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
CAS Number: 85272-31-7
UN: 3265
ADR: 8,II