Methyl-2-chloro-4,5-difluorobenzoate 98%
Supplier: Apollo Scientific
Synonyms: Methyl 2-chloro-4,5-difluorobenzoate, 2-Chloro-4,5-difluorobenzoic acid, methyl ester
PC50428-1G PC50428-5G PC50428-25G
APOSPC50428-1GEA 21.9 EUR
APOSPC50428-1G APOSPC50428-5G APOSPC50428-25G
Methyl-2-chloro-4,5-difluorobenzoate 98%
Methyl-2-chloro-4,5-difluorobenzoate
Formula: C₈H₅ClF₂O₂
MW: 206.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566827
CAS Number: 128800-36-2