1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene 97%
Supplier: Apollo Scientific
Danger
Synonyms: Methyl 1,3,3,3-tetrafluoro-2-(trifluoromethyl)prop-1-en-1-yl ether
PC5060D-25G PC5060D-5G
APOSPC5060D-25GEA 1040 EUR
APOSPC5060D-25G APOSPC5060D-5G
1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene 97%
1,1,1,3-Tetrafluoro-2-(trifluoromethyl)-4-oxapent-2-ene
Formula: C₅H₃F₇O
MW: 212.07 g/mol
Boiling Pt: 98…104 °C
Density: 1.508 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054679
CAS Number: 360-53-2
UN: 2810
ADR: 6.1,III