3-Chloro-5-nitrobenzotrifluoride 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-3-nitro-5-(trifluoromethyl)benzene
PC5062-1G PC5062-25G PC5062-5G
APOSPC5062-1GEA 21.9 EUR
APOSPC5062-1G APOSPC5062-25G APOSPC5062-5G
3-Chloro-5-nitrobenzotrifluoride 95%
3-Chloro-5-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
MW: 225.5500 g/mol
Boiling Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD03412202
CAS Number: 401-93-4