2,3,5-Trifluorobenzenesulphonyl chloride 96%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,3,5-Trifluorobenzene-1-sulphonyl chloride
PC5063-1G
PC5063-5G
APOSPC5063-1GEA
219
EUR
APOSPC5063-1G
APOSPC5063-5G
2,3,5-Trifluorobenzenesulphonyl chloride 96%
2,3,5-Trifluorobenzenesulphonyl chloride
Formula:
C₆H₂ClF₃O₂S Melting Pt: 47…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD08532462 CAS Number: 914636-99-0 UN: 3261 ADR: 8,II |