Methyl-2,6-difluorobenzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2,6-difluorobenzoate
PC5075E-100G PC5075E-25G
APOSPC5075E-100GEA 80 EUR
APOSPC5075E-100G APOSPC5075E-25G
Methyl-2,6-difluorobenzoate 97%
Methyl 2,6-difluorobenzoate
Formula: C₈H₆F₂O₂
Boiling Pt: 203…204 °C
Density: 1.281 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00051778
CAS Number: 13671-00-6