3-Fluoro-o-anisidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Fluoro-2-methoxyaniline, 3-Fluoro-2-anisidine, 2-Amino-6-fluoroanisole
PC5085-1G PC5085-25G PC5085-5G
APOSPC5085-1GEA 21.9 EUR
APOSPC5085-1G APOSPC5085-25G APOSPC5085-5G
3-Fluoro-o-anisidine 97%
3-Fluoro-o-anisidine
Formula: C₇H₈FNO
MW: 141.15 g/mol
Boiling Pt: 94 °C/10mmHg
Density: 1.172 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007759
CAS Number: 437-83-2
UN: 2810
ADR: 6.1,III