Methyl-2-fluorobenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-fluorobenzoate, 2-Fluorobenzoic acid emthyl ester
PC5088-100G PC5088-250G PC5088-25G
APOSPC5088-100GEA 43.5 EUR
APOSPC5088-100G APOSPC5088-250G APOSPC5088-25G
Methyl-2-fluorobenzoate 98%
Methyl 2-fluorobenzoate
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 109…110 °C/35mmHg
Density: 1.21 g/cm³ (25 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00017913
CAS Number: 394-35-4
EINECS: 206-894-0