6-(2-Fluorophenyl)nicotinic acid
Supplier: Apollo Scientific
PC510066-1G PC510066-5G
APOSPC510066-1GEA 429 EUR
APOSPC510066-1G APOSPC510066-5G
6-(2-Fluorophenyl)nicotinic acid
6-(2-Fluorophenyl)nicotinic acid
Formula: C₁₂H₈FNO₂
MW: 217.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410386
CAS Number: 505082-91-7