5-(4-Fluorophenyl)-3-pyridinecarbaldehyde 97%
Supplier: Apollo Scientific
Synonyms: 5-(4-Fluorophenyl)nicotinaldehyde
PC510073-1G PC510073-5G
APOSPC510073-1GEA 169 EUR
APOSPC510073-1G APOSPC510073-5G
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde 97%
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde
Formula: C₁₂H₈FNO
MW: 201.2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093924
CAS Number: 381684-96-4