6-(3,4-Difluorophenyl)-3-pyridinecarbaldehyde
Supplier: Apollo Scientific
PC510077-1G PC510077-5G
APOSPC510077-1GEA 397 EUR
APOSPC510077-1G APOSPC510077-5G
6-(3,4-Difluorophenyl)-3-pyridinecarbaldehyde
6-(3,4-Difluorophenyl)-3-pyridinecarbaldehyde
Formula: C₁₂H₇F₂NO
Storage Temperature: Ambient
CAS Number: 898404-54-1