2-Chloro-5-fluoropyrimidin-4-one 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-5-fluoro-4-pyrimidinone
PC51106-5G
APOSPC51106-5GEA 143 EUR
APOSPC51106-5G
2-Chloro-5-fluoropyrimidin-4-one 95+%
2-Chloro-5-fluoropyrimidin-4-one
Formula: C₄H₂ClFN₂O
MW: 148.5229 g/mol
Melting Pt: 176…177 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01346689
CAS Number: 155-12-4
UN: 2811
ADR: 6.1,III