2-Bromo-1-fluoro-3-iodobenzene 95%
Supplier: Apollo Scientific
Warning
PC51221-1G PC51221-250MG PC51221-5G
APOSPC51221-1GEA 77 EUR
APOSPC51221-1G APOSPC51221-250MG APOSPC51221-5G
2-Bromo-1-fluoro-3-iodobenzene 95%
2-Bromo-1-fluoro-3-iodobenzene
Formula: C₆H₃BrFI
MW: 300.8949 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09056796
CAS Number: 851368-08-6