Methyl pentafluorobenzoate 99%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl Pentafluorobenzoate
PC5123-25G PC5123-10G
APOSPC5123-25GEA 37.7 EUR
APOSPC5123-25G APOSPC5123-10G
Methyl pentafluorobenzoate 99%
Methyl pentafluorobenzoate
Formula: C₈H₃F₅O₂
MW: 226.1002 g/mol
Boiling Pt: 87…88 °C/20mmHg
Density: 1.532 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00012172
CAS Number: 36629-42-2