2,6-Difluoro-4-nitroaniline 95+%
Supplier: Apollo Scientific
Warning
PC5184-1G PC5184-5G PC5184-10G
APOSPC5184-1GEA 74.1 EUR
APOSPC5184-1G APOSPC5184-5G APOSPC5184-10G
2,6-Difluoro-4-nitroaniline 95+%
2,6-Difluoro-4-nitroaniline
Formula: C₆H₄F₂N₂O₂
MW: 174.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094274
CAS Number: 23156-27-6