2-(Trifluoromethyl)pyrazine 97%
Supplier: Apollo Scientific
PC520544-10G PC520544-5G
APOSPC520544-10GEA 745 EUR
APOSPC520544-10G APOSPC520544-5G
2-(Trifluoromethyl)pyrazine 97%
2-(Trifluoromethyl)pyrazine
Formula: C₅H₃F₃N₂
MW: 148.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797734
CAS Number: 61655-67-2