5-(Trifluoromethyl)nicotinonitrile
Supplier: Apollo Scientific
PC520548-1G PC520548-5G
APOSPC520548-1GEA 219 EUR
APOSPC520548-1G APOSPC520548-5G
5-(Trifluoromethyl)nicotinonitrile
5-(Trifluoromethyl)nicotinonitrile
Formula: C₇H₃F₃N₂
MW: 172.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368841
CAS Number: 951624-83-2