3,6-Dichloro-4-(trifluoromethyl)pyridazine
Supplier: Apollo Scientific
Warning
PC520563-5G PC520563-1G
APOSPC520563-5GEA 725 EUR
APOSPC520563-5G APOSPC520563-1G
3,6-Dichloro-4-(trifluoromethyl)pyridazine
3,6-Dichloro-4-(trifluoromethyl)pyridazine
Formula: C₅HCl₂F₃N₂
Melting Pt: 45…50 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704310
CAS Number: 1057672-68-0