1-Bromo-3-(1,1-difluoroethyl)benzene 97%
Supplier: Apollo Scientific
Warning
PC520586-1G PC520586-25G PC520586-5G
APOSPC520586-1GEA 84.1 EUR
APOSPC520586-1G APOSPC520586-25G APOSPC520586-5G
1-Bromo-3-(1,1-difluoroethyl)benzene 97%
1-Bromo-3-(1,1-difluoroethyl)benzene
Formula: C₈H₇BrF₂
MW: 221.05 g/mol
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 87.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11007736
CAS Number: 445303-70-8
UN: 3082
ADR: 9+EHS III