3-Chloro-2-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
PC520592-10G PC520592-1G PC520592-5G
APOSPC520592-10GEA 1290 EUR
APOSPC520592-10G APOSPC520592-1G APOSPC520592-5G
3-Chloro-2-(trifluoromethyl)pyridine 97%
3-Chloro-2-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD11044298
CAS Number: 749875-32-9