2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene 95%
Supplier: Apollo Scientific
PC520630-1G PC520630-5G
APOSPC520630-1GEA 176 EUR
APOSPC520630-1G APOSPC520630-5G
2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene 95%
2-(1,1-Difluoroethyl)-1-fluoro-4-(trifluoromethyl)benzene
Formula: C₉H₆F₆
MW: 228.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226715
CAS Number: 1022980-26-2