4-Fluoro-1,2-bis-(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
PC520657-25G PC520657-5G
APOSPC520657-25GEA 582 EUR
APOSPC520657-25G APOSPC520657-5G
4-Fluoro-1,2-bis-(trifluoromethyl)benzene 97%
4-Fluoro-1,2-bis-(trifluoromethyl)benzene
Formula: C₈H₃F₇
MW: 232.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100160
CAS Number: 1092349-75-1