1-(1,1-Difluoroethyl)-4-fluorobenzene 95%
Supplier: Apollo Scientific
PC520666-1G PC520666-500MG PC520666-5G
APOSPC520666-1GEA 315 EUR
APOSPC520666-1G APOSPC520666-500MG APOSPC520666-5G
1-(1,1-Difluoroethyl)-4-fluorobenzene 95%
1-(1,1-Difluoroethyl)-4-fluorobenzene
Formula: C₈H₇F₃
MW: 160.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845816
CAS Number: 55805-03-3