2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene 95%
Supplier: Apollo Scientific
PC520668-25G PC520668-5G
APOSPC520668-25GEA 861 EUR
APOSPC520668-25G APOSPC520668-5G
2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene 95%
2-Bromo-1-(1,1-difluoroethyl)-4-fluorobenzene
Formula: C₈H₆BrF₃
MW: 239.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845796
CAS Number: 1138444-86-6