3,6-Difluoro-2-methoxybenzonitrile 97%
Supplier: Apollo Scientific
PC520683-10G PC520683-1G PC520683-5G
APOSPC520683-10GEA 605 EUR
APOSPC520683-10G APOSPC520683-1G APOSPC520683-5G
3,6-Difluoro-2-methoxybenzonitrile 97%
3,6-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546978
CAS Number: 1010413-52-1