3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%
Supplier: Apollo Scientific
Danger
PC520693-1G PC520693-5G
APOSPC520693-1GEA 42.2 EUR
APOSPC520693-1G APOSPC520693-5G
3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%
3-Bromo-5-(trifluoromethyl)phenylacetic acid
Formula: C9H6BrF3O2
MW: 283.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194218
CAS Number: 1161362-01-1
UN: 3261
ADR: 8,II