2-Chloro-4-(difluoromethyl)-1-fluorobenzene 95%
Supplier: Apollo Scientific
PC520704-1G PC520704-25G PC520704-5G
APOSPC520704-1GEA 141 EUR
APOSPC520704-1G APOSPC520704-25G APOSPC520704-5G
2-Chloro-4-(difluoromethyl)-1-fluorobenzene 95%
2-Chloro-4-(difluoromethyl)-1-fluorobenzene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698545
CAS Number: 1214372-57-2