2-(Difluoromethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%
Supplier: Apollo Scientific
PC520714-1G PC520714-5G
APOSPC520714-1GEA 245 EUR
APOSPC520714-1G APOSPC520714-5G
2-(Difluoromethyl)-4-fluoro-1-(trifluoromethyl)benzene 95%
2-(Difluoromethyl)-4-fluoro-1-(trifluoromethyl)benzene
Formula: C₈H₄F₆
MW: 214.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698654
CAS Number: 1214358-19-6