2-Bromo-5-(1,1-difluoroethyl)pyridine 97%
Supplier: Apollo Scientific
PC520716-1G PC520716-5G
APOSPC520716-1GEA 279 EUR
APOSPC520716-1G APOSPC520716-5G
2-Bromo-5-(1,1-difluoroethyl)pyridine 97%
2-Bromo-5-(1,1-difluoroethyl)pyridine
Formula: C₇H₆BrF₂N
MW: 222.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16140182
CAS Number: 1211521-60-6