3,5-Dichloro-4-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
PC520719-1G PC520719-5G
APOSPC520719-1GEA 268 EUR
APOSPC520719-1G APOSPC520719-5G
3,5-Dichloro-4-(trifluoromethyl)pyridine 97%
3,5-Dichloro-4-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
Storage Temperature: Ambient
MDL Number: MFCD16140184
CAS Number: 1221272-92-9