2-(1,1-Difluoroethyl)-4-methylpyridine 97%
Supplier: Apollo Scientific
PC520764-1G PC520764-500MG PC520764-5G
APOSPC520764-1GEA 361 EUR
APOSPC520764-1G APOSPC520764-500MG APOSPC520764-5G
2-(1,1-Difluoroethyl)-4-methylpyridine 97%
2-(1,1-Difluoroethyl)-4-methylpyridine
Formula: C₈H₉F₂N
MW: 157.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22421955
CAS Number: 1401425-30-6