Potassium tris(trifluoromethanesulphonyl)methanide
Supplier: Apollo Scientific
Danger
PC52080-1G PC52080-5G
APOSPC52080-1GEA 78.3 EUR
APOSPC52080-1G APOSPC52080-5G
Potassium tris(trifluoromethanesulphonyl)methanide
Potassium tris(trifluoromethanesulphonyl)methanide
Formula: C₄F₉KO₆S₃
MW: 450.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072174
CAS Number: 114395-69-6
UN: 3263
ADR: 8,III