3,4-Difluoro-2-methylbenzotrifluoride 97%
Supplier: Apollo Scientific
PC520888-1G PC520888-5G
APOSPC520888-1GEA 306 EUR
APOSPC520888-1G APOSPC520888-5G
3,4-Difluoro-2-methylbenzotrifluoride 97%
3,4-Difluoro-2-methylbenzotrifluoride
Formula: C₈H₅F₅
MW: 196.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100534
CAS Number: 1099597-95-1