2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 95%
Supplier: Apollo Scientific
PC520909-1G PC520909-5G
APOSPC520909-1GEA 97.3 EUR
APOSPC520909-1G APOSPC520909-5G
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine 95%
2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine
Formula: C₇H₈F₃N₃
MW: 191.1537 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834983
CAS Number: 126069-70-3