1-Bromo-4-chloro-2-(2,2,2-trifluoroethyl)benzene 95%
Supplier: Apollo Scientific
PC520912-1G PC520912-5G
APOSPC520912-1GEA 260 EUR
APOSPC520912-1G APOSPC520912-5G
1-Bromo-4-chloro-2-(2,2,2-trifluoroethyl)benzene 95%
1-Bromo-4-chloro-2-(2,2,2-trifluoroethyl)benzene
Formula: C₈H₅BrClF₃
Storage Temperature: Ambient
MDL Number: MFCD11226573
CAS Number: 1099597-30-4