Nickel(II) fluoride, anhydrous 98%
Supplier: Apollo Scientific
Danger
Synonyms: Nickel difluoride
PC5210-100G PC5210-10G PC5210-25G
APOSPC5210-100GEA 319 EUR
APOSPC5210-100G APOSPC5210-10G APOSPC5210-25G
Nickel(II) fluoride, anhydrous 98%
Nickel (II) fluoride
Formula: F₂Ni
MW: 96.69 g/mol
Melting Pt: 1380 °C
Density: 4.72 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016263
CAS Number: 10028-18-9
EINECS: 233-071-3
UN: 3288
ADR: 6.1,III