2-Nitro-α,α,α-trifluorotoluene 98%
Supplier: Apollo Scientific
Danger
PC5240-25G PC5240-100G
APOSPC5240-25GEA 126 EUR
APOSPC5240-25G APOSPC5240-100G
2-Nitro-α,α,α-trifluorotoluene 98%
2-Nitro-α,α,α-trifluorotoluene
Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Boiling Pt: 104…105 °C/20mmHg
Melting Pt: 31…32 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007145
CAS Number: 384-22-5
UN: 2810
ADR: 6.1,III