5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde 95%
Supplier: Apollo Scientific
PC52471-1G PC52471-250MG
APOSPC52471-1GEA 127 EUR
APOSPC52471-1G APOSPC52471-250MG
5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde 95%
5-Chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22682805
CAS Number: 1823049-90-6